Assay Details
| Gene Symbol: snf-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064334.5 | NP_496735.1 | 1 - 2 | 244 | 89 | |
| GenBank mRNA | AY825249.1 | - | 1 - 2 | 134 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

snf-5| Gene Symbol: snf-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064334.5 | NP_496735.1 | 1 - 2 | 244 | 89 | |
| GenBank mRNA | AY825249.1 | - | 1 - 2 | 134 | 89 | |
Set Membership: |
|