Assay Details
Gene Symbol: bub-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064478.6 | NP_496879.1 | 1 - 2 | 251 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
