Assay Details
Gene Symbol: mcd-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267483.3 | NP_001254412.1 | 6 - 7 | 2690 | 75 | |
NM_001267484.3 | NP_001254413.1 | 1 - 2 | 646 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
