Assay Details
| Gene Symbol: ace-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064561.6 | NP_496962.1 | 11 - 12 | 1226 | 127 | |
| GenBank mRNA | AF025379.2 | - | 11 - 12 | 1248 | 127 | |
More Information
Set Membership: |
|

ace-4
Genomic Map