Assay Details
| Gene Symbol: ace-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064561.6 | NP_496962.1 | 4 - 5 | 584 | 71 | |
| GenBank mRNA | AF025379.2 | - | 4 - 5 | 606 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ace-4| Gene Symbol: ace-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064561.6 | NP_496962.1 | 4 - 5 | 584 | 71 | |
| GenBank mRNA | AF025379.2 | - | 4 - 5 | 606 | 71 | |
Set Membership: |
|