Assay Details
| Gene Symbol: smg-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306630.4 | NP_001293559.1 | 7 - 8 | 1641 | 98 | |
| NM_064602.6 | NP_497003.3 | 6 - 7 | 1581 | 98 | ||
More Information
Set Membership: |
|

smg-8
Genomic Map