Assay Details
| Gene Symbol: unc-52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027317.8 | NP_001022488.1 | 20 - 21 | 6846 | 78 | |
| NM_001267510.3 | NP_001254439.1 | 21 - 22 | 6257 | 78 | ||
| NM_001267511.3 | NP_001254440.1 | 19 - 20 | 6476 | 78 | ||
| NM_001267513.3 | NP_001254442.1 | 19 - 20 | 6267 | 78 | ||
| NM_001267514.4 | NP_001254443.1 | 20 - 21 | 6455 | 78 | ||
| NM_001267515.3 | NP_001254444.1 | 21 - 22 | 6737 | 78 | ||
| NM_001267516.3 | NP_001254445.1 | 21 - 22 | 6752 | 78 | ||
| NM_001267517.3 | NP_001254446.1 | 22 - 23 | 7025 | 78 | ||
| NM_001350388.3 | NP_001337294.1 | 32 | 9401 | 78 | ||
| NM_001377905.1 | NP_001364664.1 | 31 - 32 | 9294 | 78 | ||
| NM_064643.10 | NP_497044.3 | 22 - 23 | 7034 | 78 | ||
| NM_064644.6 | NP_497045.1 | 20 - 21 | 6563 | 78 | ||
| NM_064645.6 | NP_497046.1 | 22 - 23 | 7125 | 78 | ||
More Information
Set Membership: |
|

unc-52
Genomic Map