Assay Details
| Gene Symbol: unc-52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027317.8 | NP_001022488.1 | 15 - 16 | 5180 | 72 | |
| NM_001267510.3 | NP_001254439.1 | 16 - 17 | 4591 | 72 | ||
| NM_001267511.3 | NP_001254440.1 | 14 - 15 | 4810 | 72 | ||
| NM_001267515.3 | NP_001254444.1 | 16 - 17 | 5071 | 72 | ||
| NM_001267517.3 | NP_001254446.1 | 17 - 18 | 5368 | 72 | ||
| NM_001350388.3 | NP_001337294.1 | 27 | 7735 | 72 | ||
| NM_001377905.1 | NP_001364664.1 | 26 - 27 | 7628 | 72 | ||
| NM_064643.10 | NP_497044.3 | 17 - 18 | 5368 | 72 | ||
| NM_064645.6 | NP_497046.1 | 17 - 18 | 5459 | 72 | ||
More Information
Set Membership: |
|

unc-52
Genomic Map