Assay Details
| Gene Symbol: smg-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064657.3 | NP_497058.1 | 4 - 5 | 1018 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

smg-9| Gene Symbol: smg-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064657.3 | NP_497058.1 | 4 - 5 | 1018 | 73 | |
Set Membership: |
|