Assay Details
| Gene Symbol: cdh-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064674.5 | NP_497075.2 | 8 - 9 | 2686 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cdh-7| Gene Symbol: cdh-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064674.5 | NP_497075.2 | 8 - 9 | 2686 | 96 | |
Set Membership: |
|