Assay Details
| Gene Symbol: mrpl-40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064737.5 | NP_497138.1 | 3 - 4 | 324 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mrpl-40| Gene Symbol: mrpl-40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064737.5 | NP_497138.1 | 3 - 4 | 324 | 59 | |
Set Membership: |
|