Assay Details
| Gene Symbol: pals-24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064745.5 | NP_497146.2 | 10 | 922 | 104 | |
| Gene Symbol: pals-23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381682.1 | NP_001367587.1 | 9 | 908 | 104 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001381682.1 transcripts.
Set Membership: |
|

pals-24
Genomic Map