Assay Details
| Gene Symbol: klp-20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064777.6 | NP_497178.1 | 6 - 7 | 701 | 74 | |
| Gene Symbol: F40G9.17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381685.1 | NP_001367841.1 | 8 - 9 | 1067 | 74 | |
| Gene Symbol: hpo-38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001384002.1 | 8 - 9 | 1061 | 74 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001384002.1 transcripts.
Set Membership: |
|

klp-20
Genomic Map