Assay Details
Gene Symbol: klp-20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064777.6 | NP_497178.1 | 7 - 8 | 838 | 139 |
Gene Symbol: F40G9.17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381685.1 | NP_001367841.1 | 9 - 10 | 1204 | 139 |
Gene Symbol: hpo-38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001384002.1 | 9 - 10 | 1198 | 139 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001384002.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
