Assay Details
| Gene Symbol: dac-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025958.4 | NP_001021129.1 | 2 - 3 | 446 | 127 | |
| NM_001025959.5 | NP_001021130.1 | 2 - 3 | 363 | 127 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001025959.5 transcripts.
Set Membership: |
|

dac-1
Genomic Map