Assay Details
Gene Symbol: dpy-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381759.2 | NP_001367968.1 | 9 - 10 | 2250 | 110 | |
NM_001384037.1 | 13 - 14 | 3259 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
