Assay Details
| Gene Symbol: let-805 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027469.3 | NP_001022640.1 | 7 - 8 | 1792 | 86 | |
| NM_001027470.3 | NP_001022641.1 | 7 - 8 | 1792 | 86 | ||
| NM_001027472.5 | NP_001022643.1 | 7 - 8 | 1779 | 86 | ||
| GenBank mRNA | AF148953.1 | - | 7 - 8 | 1799 | 86 | |
| AF148954.1 | - | 7 - 8 | 1799 | 86 | ||
More Information
Set Membership: |
|

let-805
Genomic Map