Assay Details
| Gene Symbol: tufm-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065222.6 | NP_497623.1 | 2 - 3 | 251 | 82 | |
| GenBank mRNA | D38471.1 | - | 2 - 3 | 251 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

tufm-1| Gene Symbol: tufm-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065222.6 | NP_497623.1 | 2 - 3 | 251 | 82 | |
| GenBank mRNA | D38471.1 | - | 2 - 3 | 251 | 82 | |
Set Membership: |
|