Assay Details
| Gene Symbol: pph-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065313.6 | NP_497714.2 | 2 - 3 | 165 | 66 | |
More Information
Set Membership: | 
		   
					
						
							 | 
				
Please enter the information below and press OK to send your cart to Core Services for purchase.

pph-6| Gene Symbol: pph-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065313.6 | NP_497714.2 | 2 - 3 | 165 | 66 | |
Set Membership: | 
		   
					
						
							 |