Assay Details
Gene Symbol: mtm-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027623.6 | NP_001022794.2 | 7 - 8 | 2106 | 70 | |
NM_065365.9 | NP_497766.3 | 7 - 8 | 2094 | 70 | ||
GenBank mRNA | DQ988041.1 | - | 7 - 8 | 1955 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
