Assay Details
Gene Symbol: evl-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065464.8 | NP_497865.2 | 17 - 18 | 3452 | 151 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
