Assay Details
| Gene Symbol: ckb-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065478.4 | NP_497879.2 | 6 - 7 | 820 | 137 | |
| GenBank mRNA | AY158690.1 | - | 5 - 6 | 654 | 137 | |
More Information
Set Membership: |
|

ckb-1
Genomic Map