Assay Details
| Gene Symbol: snfc-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001384071.1 | 3 - 4 | 1010 | 58 | ||
| Gene Symbol: rnp-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001384072.1 | 3 - 4 | 1011 | 58 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001384072.1 transcripts.
Set Membership: |
|

snfc-5
Genomic Map