Assay Details
| Gene Symbol: cct-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171104.6 | NP_741117.1 | 5 - 6 | 1210 | 78 | |
| GenBank mRNA | U25698.1 | - | 5 - 6 | 1180 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cct-5| Gene Symbol: cct-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171104.6 | NP_741117.1 | 5 - 6 | 1210 | 78 | |
| GenBank mRNA | U25698.1 | - | 5 - 6 | 1180 | 78 | |
Set Membership: |
|