Assay Details
Gene Symbol: pan-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065524.5 | NP_497925.1 | 1 - 2 | 136 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
