Assay Details
Gene Symbol: pan-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065524.5 | NP_497925.1 | 4 - 5 | 666 | 147 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
