Assay Details
| Gene Symbol: mlcd-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027390.8 | NP_001022561.1 | 1 - 2 | 117 | 81 | |
| NR_002369.1 | 1 - 2 | 104 | 81 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mlcd-1| Gene Symbol: mlcd-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027390.8 | NP_001022561.1 | 1 - 2 | 117 | 81 | |
| NR_002369.1 | 1 - 2 | 104 | 81 | |||
Set Membership: |
|