Assay Details
| Gene Symbol: sel-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001377920.3 | NP_001364559.1 | 13 - 14 | 3328 | 62 | |
| NM_065538.6 | NP_497939.2 | 12 - 13 | 3193 | 62 | ||
More Information
Set Membership: |
|

sel-2
Genomic Map