Assay Details
| Gene Symbol: sel-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001377920.3 | NP_001364559.1 | 23 - 24 | 5399 | 79 | |
| NM_065538.6 | NP_497939.2 | 22 - 23 | 5264 | 79 | ||
More Information
Set Membership: |
|

sel-2
Genomic Map