Assay Details
| Gene Symbol: pfas-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065541.8 | NP_497942.2 | 7 - 8 | 1143 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pfas-1| Gene Symbol: pfas-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065541.8 | NP_497942.2 | 7 - 8 | 1143 | 78 | |
Set Membership: |
|