Assay Details
Gene Symbol: clp-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065563.4 | NP_497964.1 | 2 - 3 | 264 | 130 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
