Assay Details
| Gene Symbol: emb-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065568.4 | NP_497969.1 | 6 - 7 | 2194 | 109 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

emb-5| Gene Symbol: emb-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065568.4 | NP_497969.1 | 6 - 7 | 2194 | 109 | |
Set Membership: |
|