Assay Details
| Gene Symbol: ula-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001361710.4 | NP_001348740.1 | 3 - 4 | 543 | 118 | |
| NM_065636.5 | NP_498037.2 | 3 - 4 | 541 | 118 | ||
| Gene Symbol: mmab-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001384085.1 | 7 - 8 | 1572 | 118 | ||
More Information
Set Membership: |
|

ula-1
Genomic Map