Assay Details
| Gene Symbol: larp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047403.5 | NP_001040868.3 | 10 - 11 | 2742 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

larp-1| Gene Symbol: larp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047403.5 | NP_001040868.3 | 10 - 11 | 2742 | 79 | |
Set Membership: |
|