Assay Details
| Gene Symbol: larp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047402.3 | NP_001040867.2 | 5 - 6 | 1546 | 118 | |
| NM_001047403.5 | NP_001040868.3 | 8 - 9 | 2365 | 118 | ||
More Information
Set Membership: |
|

larp-1
Genomic Map