Assay Details
| Gene Symbol: ulp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065694.8 | NP_498095.3 | 5 - 6 | 927 | 102 | |
| GenBank mRNA | AB095020.1 | - | 5 - 6 | 867 | 102 | |
| Gene Symbol: T10F2.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001382862.1 | 5 - 6 | 888 | 102 | ||
| Gene Symbol: prp-19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001382863.1 | 8 - 9 | 2970 | 102 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001382862.1 transcripts.
Set Membership: |
|

ulp-1
Genomic Map