Assay Details
| Gene Symbol: hap-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065720.4 | NP_498121.1 | 2 - 3 | 194 | 69 | |
| GenBank mRNA | AY052772.1 | - | 2 - 3 | 194 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

hap-1| Gene Symbol: hap-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065720.4 | NP_498121.1 | 2 - 3 | 194 | 69 | |
| GenBank mRNA | AY052772.1 | - | 2 - 3 | 194 | 69 | |
Set Membership: |
|