Assay Details
| Gene Symbol: gro-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065721.7 | NP_498122.2 | 2 - 3 | 335 | 77 | |
| GenBank mRNA | AY052773.1 | - | 2 - 3 | 335 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

gro-1| Gene Symbol: gro-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065721.7 | NP_498122.2 | 2 - 3 | 335 | 77 | |
| GenBank mRNA | AY052773.1 | - | 2 - 3 | 335 | 77 | |
Set Membership: |
|