Assay Details
Gene Symbol: thoc-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267997.2 | NP_001254926.1 | 3 - 4 | 589 | 63 | |
NM_001267998.4 | NP_001254927.1 | 3 - 4 | 623 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
