Assay Details
| Gene Symbol: rgs-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065828.8 | NP_498229.3 | 4 - 5 | 745 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rgs-5| Gene Symbol: rgs-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065828.8 | NP_498229.3 | 4 - 5 | 745 | 89 | |
Set Membership: |
|