Assay Details
| Gene Symbol: rgs-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129200.4 | NP_001122672.1 | 5 - 6 | 1364 | 82 | |
| NM_065828.8 | NP_498229.3 | 6 - 7 | 1588 | 82 | ||
More Information
Set Membership: |
|

rgs-5
Genomic Map