Assay Details
Gene Symbol: nmt-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065925.5 | NP_498326.1 | 1 - 2 | 114 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
