Assay Details
Gene Symbol: rbf-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027395.5 | NP_001022566.1 | 15 - 16 | 2713 | 117 | |
NM_001027396.5 | NP_001022567.1 | 15 - 16 | 2515 | 117 | ||
NM_001379772.1 | NP_001367825.1 | 17 - 18 | 3226 | 117 | ||
NM_066066.7 | NP_498467.3 | 17 - 18 | 3189 | 117 | ||
GenBank mRNA | AF399852.1 | - | 14 - 15 | 2493 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
