Assay Details
Gene Symbol: rbf-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027395.5 | NP_001022566.1 | 17 - 18 | 2970 | 61 | |
NM_001027396.5 | NP_001022567.1 | 17 - 18 | 2772 | 61 | ||
NM_001379772.1 | NP_001367825.1 | 19 - 20 | 3483 | 61 | ||
NM_066066.7 | NP_498467.3 | 19 - 20 | 3446 | 61 | ||
GenBank mRNA | AF399852.1 | - | 16 - 17 | 2750 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
