Assay Details
Gene Symbol: ten-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027552.5 | NP_001022723.2 | 3 - 4 | 318 | 70 | |
NM_001268011.3 | NP_001254940.1 | 6 - 7 | 712 | 70 | ||
NM_001268012.3 | NP_001254941.1 | 4 - 5 | 490 | 70 | ||
NM_001379777.2 | NP_001367791.1 | 3 - 4 | 324 | 70 | ||
GenBank mRNA | AB206835.1 | - | 3 - 4 | 318 | 70 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001379777.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
