Assay Details
| Gene Symbol: ogt-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047395.4 | NP_001040860.1 | 12 - 13 | 3317 | 75 | |
| NM_001047396.5 | NP_001040861.1 | 8 - 9 | 2749 | 75 | ||
More Information
Set Membership: |
|

ogt-1
Genomic Map