Assay Details
| Gene Symbol: ogt-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047395.4 | NP_001040860.1 | 7 - 8 | 1305 | 110 | |
| NM_001047396.5 | NP_001040861.1 | 3 - 4 | 737 | 110 | ||
More Information
Set Membership: |
|

ogt-1
Genomic Map