Assay Details
| Gene Symbol: rnp-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066164.8 | NP_498565.2 | 3 - 4 | 405 | 94 | |
| Gene Symbol: syf-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001379785.1 | NP_001367336.1 | 5 - 6 | 1274 | 94 | |
More Information
Set Membership: |
|

rnp-7
Genomic Map