Assay Details
| Gene Symbol: fbn-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066269.6 | NP_498670.2 | 15 - 16 | 6009 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

fbn-1| Gene Symbol: fbn-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066269.6 | NP_498670.2 | 15 - 16 | 6009 | 62 | |
Set Membership: |
|