Assay Details
| Gene Symbol: lido-18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001379800.2 | NP_001367413.1 | 7 - 8 | 3407 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

lido-18| Gene Symbol: lido-18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001379800.2 | NP_001367413.1 | 7 - 8 | 3407 | 74 | |
Set Membership: |
|